siRNA Details
Gene Symbol: Slc25a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053515.1 | NP_445967.1 | 2 | 685 | ||
GenBank mRNA | BC060533.1 | - | 2 | 689 | 6922056 | |
D12770.1 | - | 2 | 652 | |||
FQ213027.1 | - | 2 | 682 | |||
FQ213086.1 | - | 2 | 682 | |||
FQ213612.1 | - | 2 | 711 | |||
FQ214384.1 | - | 2 | 681 | |||
FQ220525.1 | - | 2 | 680 | |||
FQ224814.1 | - | 2 | 682 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007450, AK051172, BC003791, BC026925, AK078077, and others.
Gene Ontology Categories:
