siRNA Details
Gene Symbol: Ngb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033359.3 | NP_203523.2 | 2 | 595 | ||
XM_008764864.1 | XP_008763086.1 | 2 | 1276 | |||
GenBank mRNA | AB056655.1 | - | 2 | 167 | ||
AF333245.1 | - | 2 | 186 | |||
AF334379.1 | - | 2 | 186 | |||
AY066001.1 | - | 2 | 395 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
