siRNA Details
Gene Symbol: Ngb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033359.3 | NP_203523.2 | 2, 3 | 603 | ||
XM_008764864.1 | XP_008763086.1 | 2, 3 | 1284 | |||
GenBank mRNA | AB056655.1 | - | 2, 3 | 175 | ||
AF333245.1 | - | 2, 3 | 194 | |||
AF334379.1 | - | 2, 3 | 194 | |||
AY066001.1 | - | 2, 3 | 403 |
