siRNA Details
Gene Symbol: Nol3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053516.2 | NP_445968.2 | 1 | 126 | ||
XM_006255539.2 | XP_006255601.1 | 2 | 257 | |||
XM_006255542.2 | XP_006255604.1 | 2 | 257 | |||
XM_008772540.1 | XP_008770762.1 | 2 | 738 | |||
GenBank mRNA | U40627.1 | - | 1 | 126 |

More Information
Other Species Targeted

Gene Ontology Categories:
