siRNA Details
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164060.1 | NP_001157532.1 | 10 | 1474 | ||
NM_053517.2 | NP_445969.2 | 11 | 1041 | |||
GenBank mRNA | BC088298.1 | - | 11 | 1041 | 7323045 | |
CB813924.1 | - | 4 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
