siRNA Details
Gene Symbol: Prkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276715.1 | NP_001263644.1 | 15 | 2074 | ||
XM_006240074.2 | XP_006240136.1 | 14 | 2332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
