siRNA Details
Gene Symbol: Slc5a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053521.1 | NP_445973.1 | 5 | 686 | ||
XM_006244251.2 | XP_006244313.1 | 5 | 789 | |||
XM_006244253.2 | XP_006244315.1 | 3 | 333 | |||
XM_008766774.1 | XP_008764996.1 | 5 | 837 | |||
XM_008766775.1 | XP_008764997.1 | 5 | 786 | |||
GenBank mRNA | AB030947.1 | - | 5 | 686 |
