siRNA Details
Gene Symbol: Rhoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053522.1 | NP_445974.1 | 3 | 614 | ||
GenBank mRNA | AB031482.1 | - | 3 | 614 | ||
BC061760.1 | - | 3 | 604 | 5621223 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rhoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053522.1 | NP_445974.1 | 3 | 614 | ||
GenBank mRNA | AB031482.1 | - | 3 | 614 | ||
BC061760.1 | - | 3 | 604 | 5621223 |