siRNA Details
Gene Symbol: Rhoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053522.1 | NP_445974.1 | 3 | 594 | ||
GenBank mRNA | AB031482.1 | - | 3 | 594 | ||
BC061760.1 | - | 3 | 584 | 5621223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
