siRNA Details
Gene Symbol: Vamp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053531.1 | NP_445983.1 | 5 | 466 | ||
XM_006249016.2 | XP_006249078.1 | 6 | 619 | |||
GenBank mRNA | AF281632.1 | - | 5 | 466 | ||
FQ228923.1 | - | 6 | 579 | |||
FQ230649.1 | - | 5 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
