siRNA Details
Gene Symbol: Klf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053536.1 | NP_445988.1 | 3 | 1224 | ||
XM_006236882.2 | XP_006236944.1 | 2 | 1024 | |||
XM_006236884.2 | XP_006236946.1 | 2 | 1220 | |||
XM_008763135.1 | XP_008761357.1 | 2 | 1361 | |||
XM_008763136.1 | XP_008761358.1 | 2 | 2183 | |||
GenBank mRNA | AB020759.1 | - | 3 | 1224 | ||
AB102791.1 | - | 1 | 946 | |||
AB102792.1 | - | 1 | 946 | |||
BC089782.1 | - | 2 | 1123 | 7325036 |

More Information
Other Species Targeted

Gene Ontology Categories:
