siRNA Details
Gene Symbol: Sfrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053544.1 | NP_445996.1 | 3, 4 | 684 | ||
GenBank mRNA | AF012891.1 | - | 3, 4 | 684 | ||
AF140346.1 | - | 3, 4 | 835 | |||
AF220608.1 | - | 932 | ||||
FQ229224.1 | - | 2, 3 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
