siRNA Details
Gene Symbol: Cx3cl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134455.1 | NP_604450.1 | 3 | 434 | ||
XM_008772329.1 | XP_008770551.1 | 4 | 1171 | |||
GenBank mRNA | AF030358.2 | - | 3 | 434 | ||
BC070938.1 | - | 3 | 435 | 7099250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
