siRNA Details
Gene Symbol: Opn1mw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053548.1 | NP_446000.1 | 6 | 1017 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
