siRNA Details
Gene Symbol: Vegfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053549.1 | NP_446001.1 | 3, 4 | 552 | ||
XM_006230910.2 | XP_006230972.1 | 3, 4 | 649 | |||
XM_006230911.2 | XP_006230973.1 | 3, 4 | 650 | |||
XM_006230912.2 | XP_006230974.1 | 3, 4 | 640 | |||
GenBank mRNA | AF022952.1 | - | 1, 2 | 205 | ||
AF032925.1 | - | 1, 2 | 148 | |||
BC076395.1 | - | 3, 4 | 552 | 7109119 |

More Information
Other Species Targeted

Gene Ontology Categories:
