siRNA Details
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048180.1 | NP_001041645.1 | 6, 7 | 1734 | ||
NM_134456.3 | NP_604451.2 | 7, 8 | 2034 | |||
XM_006230351.2 | XP_006230413.1 | 6, 7 | 3595 | |||
XM_006230352.2 | XP_006230414.1 | 6, 7 | 3595 | |||
XM_006230353.2 | XP_006230415.1 | 7, 8 | 2009 | |||
XM_006230354.2 | XP_006230416.1 | 7, 8 | 2915 | |||
XM_006230355.2 | XP_006230417.1 | 6, 7 | 3594 | |||
GenBank mRNA | AF047577.1 | - | 6, 7 | 1734 | ||
U57391.1 | - | 7, 8 | 2077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
