siRNA Details
Gene Symbol: Prmt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053557.1 | NP_446009.1 | 7 | 749 | ||
GenBank mRNA | AF059530.1 | - | 7 | 749 |

More Information
Other Species Targeted

Gene Ontology Categories:
