siRNA Details
Gene Symbol: Chi3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053560.1 | NP_446012.1 | 6, 7 | 650 | ||
XM_006249892.2 | XP_006249954.2 | 6, 7 | 704 | |||
XM_008769574.1 | XP_008767796.1 | 6, 7 | 671 | |||
GenBank mRNA | AF062038.1 | - | 4, 5 | 531 | ||
BC091365.1 | - | 6, 7 | 650 | 7319171 | ||
EF467168.1 | - | 4, 5 | 531 | |||
FQ232458.1 | - | 3, 4 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
