siRNA Details
Gene Symbol: Rpe65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053562.3 | NP_446014.2 | 13 | 1439 | ||
GenBank mRNA | AF035673.1 | - | 13 | 1441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
