siRNA Details
Gene Symbol: Socs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053565.1 | NP_446017.1 | 1 | 595 | ||
XM_008768398.1 | XP_008766620.1 | 2 | 963 | |||
GenBank mRNA | AF075383.1 | - | 1 | 595 |

More Information
Other Species Targeted

Gene Ontology Categories:
