siRNA Details
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053571.1 | NP_446023.1 | 3 | 460 | ||
XM_008769682.1 | XP_008767904.1 | 9 | 3992 | |||
XM_008769683.1 | XP_008767905.1 | 7 | 3700 | |||
XM_008769684.1 | XP_008767906.1 | 4 | 1224 | |||
XM_008769685.1 | XP_008767907.1 | 6 | 2977 | |||
XM_008769686.1 | XP_008767908.1 | 2 | 254 | |||
GenBank mRNA | AB060653.1 | - | 3 | 460 |
