siRNA Details
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053571.1 | NP_446023.1 | 20 | 2666 | ||
XM_008769682.1 | XP_008767904.1 | 26 | 6195 | |||
XM_008769683.1 | XP_008767905.1 | 24 | 5903 | |||
XM_008769684.1 | XP_008767906.1 | 21 | 3427 | |||
XM_008769685.1 | XP_008767907.1 | 23 | 5180 | |||
XM_008769686.1 | XP_008767908.1 | 19 | 2464 | |||
GenBank mRNA | AB060653.1 | - | 20 | 2666 |
