siRNA Details
Gene Symbol: Olfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053573.1 | NP_446025.1 | 3, 4 | 746 | ||
XM_006233875.2 | XP_006233937.1 | 3, 4 | 706 | |||
GenBank mRNA | U03416.1 | - | 3, 4 | 608 | ||
U03417.1 | - | 3, 4 | 746 |

More Information
Other Species Targeted

Gene Ontology Categories:
