siRNA Details
Gene Symbol: Cabin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053575.2 | NP_446027.2 | 19, 20 | 3072 | ||
GenBank mRNA | AF061947.1 | - | 19, 20 | 3078 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cabin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053575.2 | NP_446027.2 | 19, 20 | 3072 | ||
GenBank mRNA | AF061947.1 | - | 19, 20 | 3078 |