siRNA Details
Gene Symbol: Prdx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053576.2 | NP_446028.1 | 2 | 216 | ||
GenBank mRNA | AF014009.1 | - | 2 | 216 | ||
FQ218168.1 | - | 2 | 245 | |||
Y17295.1 | - | 230 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177256, NM_007453, AK157820, AK161395, AK172888, and others.
Gene Ontology Categories:
