siRNA Details
Gene Symbol: Atp6v0e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053578.3 | NP_446030.3 | 2 | 243 | ||
GenBank mRNA | AB037248.1 | - | 2 | 243 | ||
AB051300.1 | - | 1 | 1385 | |||
FQ226644.1 | - | 2 | 206 | |||
FQ226841.1 | - | 2 | 213 | |||
FQ228396.1 | - | 2 | 200 | |||
FQ233078.1 | - | 2 | 202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
