siRNA Details
Gene Symbol: Slc27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053580.2 | NP_446032.2 | 4, 5 | 899 | ||
XM_006252953.2 | XP_006253015.1 | 4, 5 | 937 | |||
XM_006252954.1 | XP_006253016.1 | 4, 5 | 885 | |||
XM_008771156.1 | XP_008769378.1 | 4, 5 | 824 | |||
XM_008771157.1 | XP_008769379.1 | 4, 5 | 983 | |||
GenBank mRNA | BC074014.1 | - | 4, 5 | 899 | 7104661 | |
U89529.1 | - | 4, 5 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
