siRNA Details
Gene Symbol: Madd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053585.1 | NP_446037.1 | 4 | 1062 | ||
GenBank mRNA | U72995.1 | - | 4 | 1062 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Madd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053585.1 | NP_446037.1 | 4 | 1062 | ||
GenBank mRNA | U72995.1 | - | 4 | 1062 |