siRNA Details
Gene Symbol: S100a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053587.1 | NP_446039.1 | 1 | 152 | ||
XM_006232780.1 | XP_006232842.1 | 2 | 233 | |||
GenBank mRNA | AB118215.1 | - | 1 | 116 | ||
L18948.1 | - | 1 | 152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
