siRNA Details
Gene Symbol: Rnf138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053588.2 | NP_446040.1 | 349 | |||
GenBank mRNA | AF315468.1 | - | 310 | |||
BC061821.1 | - | 349 | 5599087 | |||
FQ220857.1 | - | 645 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_207623, NM_019706, AB025011, AB025010, BC003712, and others.
Gene Ontology Categories:
