siRNA Details
Gene Symbol: Psmb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053590.1 | NP_446042.1 | 3 | 305 | ||
XM_006227981.1 | XP_006228043.1 | 5 | 560 | |||
GenBank mRNA | BC058455.1 | - | 3 | 317 | 6922317 | |
FQ211516.1 | - | 3 | 319 | |||
FQ214442.1 | - | 3 | 320 | |||
FQ214726.1 | - | 5 | 560 | |||
FQ218426.1 | - | 3 | 290 | |||
FQ225328.1 | - | 3 | 320 | |||
FQ229756.1 | - | 3 | 320 | |||
FQ230094.1 | - | 3 | 320 | |||
X52783.1 | - | 3 | 305 |

More Information
Other Species Targeted

Gene Ontology Categories:
