siRNA Details
Gene Symbol: Ptprr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113390.1 | NP_001106861.1 | 8 | 1098 | ||
NM_053594.2 | NP_446046.2 | 1807 | ||||
GenBank mRNA | BC089186.1 | - | 4 | 589 | 7315399 | |
D38292.1 | - | 1810 | ||||
D64050.1 | - | 8 | 1126 | |||
U14914.1 | - | 1154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
