siRNA Details
Gene Symbol: Nrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271118.1 | NP_001258047.1 | 6, 7 | 1733 | ||
NM_001271119.1 | NP_001258048.1 | 5, 6 | 1718 | |||
NM_001271120.1 | NP_001258049.1 | 5, 6 | 1718 | |||
NM_001271121.1 | NP_001258050.1 | 7, 8 | 1162 | |||
NM_001271122.1 | NP_001258051.1 | 8, 9 | 1186 | |||
NM_001271124.1 | NP_001258053.1 | 9, 10 | 1273 | |||
NM_001271125.1 | NP_001258054.1 | 10, 11 | 1342 | |||
NM_001271126.1 | NP_001258055.1 | 10, 11 | 1288 | |||
NM_001271127.1 | NP_001258056.1 | 10, 11 | 1342 | |||
NM_001271128.1 | NP_001258057.1 | 9, 10 | 1273 | |||
NM_001271129.1 | NP_001258058.1 | 9, 10 | 1273 | |||
NM_001271130.1 | NP_001258059.1 | 10, 11 | 1358 | |||
NM_031588.2 | NP_113776.2 | 9, 10 | 1264 | |||
GenBank mRNA | AF194438.1 | - | 6, 7 | 1747 | ||
AF194439.1 | - | 5, 6 | 1466 | |||
AF194440.1 | - | 5, 6 | 541 | |||
AF194993.1 | - | 10, 11 | 1358 | |||
AY973241.1 | - | 8, 9 | 678 | |||
AY973243.1 | - | 7, 8 | 656 | |||
AY973244.1 | - | 8, 9 | 644 | |||
AY973245.1 | - | 10, 11 | 1009 | |||
AY995221.1 | - | 10, 11 | 944 | |||
AY995222.1 | - | 7, 8 | 818 | |||
AY995223.1 | - | 8, 9 | 842 | |||
M92430.1 | - | 9, 10 | 973 | |||
U02316.1 | - | 9, 10 | 1273 | |||
U02317.1 | - | 9, 10 | 1273 | |||
U02318.1 | - | 9, 10 | 1264 | |||
U02319.1 | - | 9, 10 | 1273 | |||
U02321.1 | - | 9, 10 | 1264 | |||
U02322.1 | - | 10, 11 | 1342 | |||
U02323.1 | - | 9, 10 | 1273 | |||
U02324.1 | - | 9, 10 | 973 |

More Information
Other Species Targeted

Gene Ontology Categories:
