siRNA Details
Gene Symbol: Hspb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053612.2 | NP_446064.1 | 2 | 775 | ||
GenBank mRNA | AF314540.1 | - | 2 | 457 | ||
BC061748.1 | - | 2 | 775 | 5599613 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
