siRNA Details
Gene Symbol: Lenep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053614.1 | NP_446066.1 | 2 | 479 | ||
GenBank mRNA | U15149.1 | - | 2 | 479 |
Gene Symbol: Flad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110138.1 | NP_001103608.1 | 8 | 2736 | ||
GenBank mRNA | FQ218677.1 | - | 9 | 1900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
