siRNA Details
Gene Symbol: Sep15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133297.2 | NP_579831.2 | 4 | 599 | ||
GenBank mRNA | AF390544.1 | - | 4 | 323 | ||
BC060547.1 | - | 4 | 348 | 6921948 | ||
CK596377.1 | - | 4 | 614 | |||
DY561321.1 | - | 4 | 337 | |||
FQ209574.1 | - | 4 | 334 | |||
FQ212727.1 | - | 4 | 367 | |||
FQ213177.1 | - | 4 | 348 | |||
FQ213838.1 | - | 4 | 334 | |||
FQ214634.1 | - | 4 | 344 | |||
FQ215362.1 | - | 4 | 334 | |||
FQ215992.1 | - | 4 | 357 | |||
FQ216831.1 | - | 4 | 334 | |||
FQ219237.1 | - | 4 | 355 | |||
FQ219570.1 | - | 4 | 334 | |||
FQ219725.1 | - | 4 | 327 | |||
FQ220311.1 | - | 4 | 333 | |||
FQ220593.1 | - | 4 | 339 | |||
FQ220877.1 | - | 4 | 347 | |||
FQ229133.1 | - | 4 | 340 | |||
FQ230131.1 | - | 4 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
