siRNA Details
Gene Symbol: Bbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053618.1 | NP_446070.1 | 14 | 1771 | ||
XM_006255131.2 | XP_006255193.1 | 14 | 1930 | |||
XM_008772263.1 | XP_008770485.1 | 14 | 1698 | |||
GenBank mRNA | AF342738.1 | - | 14 | 1771 | ||
FQ222286.1 | - | 8 | 1202 |
