siRNA Details
Gene Symbol: Pecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133299.2 | NP_579833.1 | 3, 4 | 524 | ||
XM_008767180.1 | XP_008765402.1 | 3, 4 | 530 | |||
GenBank mRNA | AF021854.1 | - | 3, 4 | 525 | ||
AF099742.1 | - | 3, 4 | 444 | |||
BC060546.1 | - | 3, 4 | 518 | 6889831 |
