siRNA Details
Gene Symbol: Magi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053621.1 | NP_446073.1 | 8 | 1312 | ||
GenBank mRNA | AF034863.1 | - | 8 | 1312 | ||
AF130819.1 | - | 7 | 1099 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001170745, NM_001170746, NM_015823, AK122349, AK147530, and others.
Gene Ontology Categories:
