siRNA Details
Gene Symbol: Banf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053631.3 | NP_446083.1 | 3 | 691 | ||
XM_006230630.2 | XP_006230692.1 | 3 | 704 | |||
XM_006230631.1 | XP_006230693.1 | 3 | 728 | |||
XM_006230632.2 | XP_006230694.1 | 3 | 701 | |||
GenBank mRNA | AB024333.1 | - | 3 | 688 | ||
BC084726.1 | - | 3 | 662 | 7320690 | ||
FQ224698.1 | - | 3 | 733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
