siRNA Details
Gene Symbol: Sec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276417.1 | NP_001263346.1 | 11 | 5142 | ||
XM_006233662.2 | XP_006233724.1 | 11 | 5166 | |||
XM_006233663.2 | XP_006233725.1 | 11 | 4969 | |||
XM_006233664.2 | XP_006233726.1 | 11 | 6052 | |||
XM_006233666.2 | XP_006233728.1 | 12 | 4476 | |||
GenBank mRNA | CB697736.1 | - | 2 | 228 |

More Information
Other Species Targeted

Gene Ontology Categories:
