siRNA Details
Gene Symbol: Uchl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110165.1 | NP_001103635.1 | 8 | 643 | ||
GenBank mRNA | AB043959.1 | - | 8 | 568 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Uchl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110165.1 | NP_001103635.1 | 8 | 643 | ||
GenBank mRNA | AB043959.1 | - | 8 | 568 |