siRNA Details
Gene Symbol: Cdh23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053644.1 | NP_446096.1 | 43 | 6369 | ||
GenBank mRNA | AB053447.1 | - | 43 | 6369 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdh23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053644.1 | NP_446096.1 | 43 | 6369 | ||
GenBank mRNA | AB053447.1 | - | 43 | 6369 |