siRNA Details
Gene Symbol: Cdh23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053644.1 | NP_446096.1 | 46 | 7119 | ||
GenBank mRNA | AB053447.1 | - | 46 | 7119 |

More Information
Other Species Targeted

Gene Ontology Categories:
