siRNA Details
Gene Symbol: Cxcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053647.1 | NP_446099.1 | 3 | 288 | ||
GenBank mRNA | AB060092.1 | - | 3 | 288 | ||
S77604.1 | - | 3 | 243 | |||
U45965.1 | - | 3 | 251 | |||
X65647.1 | - | 3 | 228 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
