siRNA Details
Gene Symbol: Kremen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053649.1 | NP_446101.1 | 9 | 1370 | ||
GenBank mRNA | AB065090.1 | - | 9 | 1370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
