siRNA Details
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053650.1 | NP_446102.1 | 3, 4 | 416 | ||
XM_006253120.2 | XP_006253182.1 | 3, 4 | 452 | |||
GenBank mRNA | AF002281.1 | - | 3, 4 | 434 | ||
FQ215471.1 | - | 3, 4 | 434 |
