siRNA Details
Gene Symbol: Sardh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053664.2 | NP_446116.1 | 9 | 1301 | ||
XM_006233731.2 | XP_006233793.1 | 10 | 1323 | |||
XM_006233732.2 | XP_006233794.1 | 10 | 1759 | |||
XM_006233733.2 | XP_006233795.1 | 10 | 1325 | |||
GenBank mRNA | AF067650.1 | - | 9 | 1336 | ||
FQ218777.1 | - | 9 | 1597 |
