siRNA Details
Gene Symbol: Crcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053670.3 | NP_446122.1 | 4 | 183 | ||
GenBank mRNA | AF103950.1 | - | 2 | 98 | ||
AF440799.2 | - | 4 | 183 | |||
BC059117.1 | - | 4 | 287 | 6891094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
