siRNA Details
Gene Symbol: Crcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053670.3 | NP_446122.1 | 3, 4 | 156 | ||
GenBank mRNA | AF103950.1 | - | 1, 2 | 71 | ||
AF440799.2 | - | 3, 4 | 156 | |||
BC059117.1 | - | 3, 4 | 260 | 6891094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
